ADV371425663 AChemBlock

trans-1-Boc-4-fluoro-3-hydroxypiperidine

  • MDL number MFCD18425630
  • Cas Number : 955028-82-7
  • Molecular Weight : 219.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-1-boc-4-fluoro-3-hydroxypiperidine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3593
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart