ADV371425664 AChemBlock

Trans-tert-Butyl 3-fluoro-4-hydroxypyrrolidine-1-carboxylate

  • MDL number MFCD18791200
  • Cas Number : 869481-93-6
  • Molecular Weight : 205.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid/light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-tert-butyl-3-fluoro-4-hydroxypyrrolidine-1-carboxylate.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3594
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart