ADV371425665 AChemBlock

trans-4-Amino-1-Boc-3-hydroxypiperidine

  • MDL number MFCD18425632
  • Cas Number : 443955-98-4
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid/yellow liquid/colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-4-amino-1-boc-3-hydroxypiperidine.html

Safety Information

signalword Danger
pcodes P261;P264;P270;P271;P280;P301+310;P302+352;P304+340;P305+351+338;P310;P322;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3595
pictograms GHS06; GHS05
hstatements H301;H312;H332;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart