ADV371425678 AChemBlock

4-Aminomethyl-2-methoxypyridine

  • MDL number MFCD09054679
  • Cas Number : 148900-69-0
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellowish liquid/colorless liquid or light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-aminomethyl-2-methoxypyridine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3608
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart