ADV371425679 AChemBlock

4-Aminomethyl-2-piperidone hydrochloride

  • MDL number MFCD20809862
  • Cas Number : 1400764-40-0
  • Molecular Weight : 164.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form white crystalline solid/Gray solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-aminomethyl-2-piperidone-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3610
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart