ADV371425691 AChemBlock

3-Aminocyclobutanol hydrochloride

  • MDL number MFCD12547215
  • Cas Number : 1036260-25-9
  • Molecular Weight : 123.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow liquid/pale yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-aminocyclobutanol-hydrochloride.html

Safety Information

signalword Warning
pcodes P210;P233;P240;P241;P242;P243;P264;P270;P280;P301;P301+312;P303+361+353;P305+351+338;P337+313;P370+378;P403+235
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3623
pictograms GHS02; GHS07
hstatements H226;H302;H319
Availability details for
Quantity
  • Availability
  • Add to Cart