ADV371425699 AChemBlock

4-Fluoro-7-azaindole

  • MDL number MFCD08272231
  • Cas Number : 640735-23-5
  • Molecular Weight : 136.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-fluoro-7-azaindole.html

Safety Information

signalword Warning
pcodes P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3632
pictograms GHS07
hstatements H302;H319
Availability details for
Quantity
  • Availability
  • Add to Cart