ADV371425706 AChemBlock

4-Fluoro-1-(triisopropylsilanyl)-7-azaindole

  • MDL number MFCD11616419
  • Cas Number : 640735-25-7
  • Molecular Weight : 292.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-fluoro-1-triisopropylsilanyl-7-azaindole.html

Safety Information

signalword Warning
pcodes P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3640
pictograms GHS07
hstatements H303;H319
Availability details for
Quantity
  • Availability
  • Add to Cart