ADV371425716 AChemBlock

cis-1-Boc-3-amino-4-fluoropyrrolidine

  • MDL number MFCD18072719
  • Cas Number : 1431720-86-3
  • Molecular Weight : 204.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form color liquid or white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/cis-1-boc-3-amino-4-fluoropyrrolidine.html

Safety Information

signalword Danger
pcodes P261;P264;P270;P271;P280;P301+312;P302+352;P304+340;P305+351+338;P310;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3652
pictograms GHS05; GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart