ADV371425717 AChemBlock

5-Bromo-2-chloro-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD09909707
  • Cas Number : 1060816-58-1
  • Molecular Weight : 232.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-chloro-7h-pyrrolo-2-3-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3653
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart