ADV371425719 AChemBlock

Ethyl 1H-pyrazole-3-carboxylate

  • MDL number MFCD00159643
  • Cas Number : 5932-27-4
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form white powder
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1h-pyrazole-3-carboxylate.html

Safety Information

signalword Warning
pcodes P210;P261;P264;P271;P280;P302+352;P304+340;P305+351+338;P337+313;P370+378;P403+235
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3655
pictograms GHS07
hstatements H227;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart