ADV371425731 AChemBlock

7-Acetyl-1H-indazole

  • MDL number MFCD11869765
  • Cas Number : 1159511-22-4
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-acetyl-1h-indazole.html

Safety Information

signalword Warning
pcodes P264;P280;P305+351+338;P337+313
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3670
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart