ADV371425733 AChemBlock

1-Triisopropylsilany-5-(Boc-amino)-7-azaindole

  • MDL number MFCD16039205
  • Cas Number : 651744-43-3
  • Molecular Weight : 389.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-triisopropylsilany-5-boc-amino-7-azaindole.html

Safety Information

signalword Danger
pcodes P271;P280;P310;P301+312;P302+352;P304+340;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3673
pictograms GHS05; GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart