ADV371425745 AChemBlock

6-Chloro-1H-pyrazolo[4,3-b]pyridine

  • MDL number MFCD16659575
  • Cas Number : 1260670-01-6
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-1h-pyrazolo-4-3-b-pyridine.html

Safety Information

signalword Warning
pcodes P210;P261;P264;P271;P280;P302+352;P304+340;P305+351+338;P337+313;P370+378;P403+233;P403+235
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3694
pictograms GHS07
hstatements H227;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart