ADV371425746 AChemBlock

Ethyl 6-Chloro-1H-pyrazolo[4,3-b]pyridine-3-carboxylate

  • MDL number MFCD21642076
  • Cas Number : 1363380-62-4
  • Molecular Weight : 225.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-6-chloro-1h-pyrazolo-4-3-b-pyridine-3-carboxylate.html

Safety Information

signalword Warning
pcodes P210;P261;P264;P271;P280;P302+352;P304+340;P305+351+338;P337+313;P370+378;P403+233;P403+235
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3695
pictograms GHS07
hstatements H227;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart