ADV371425748 AChemBlock

3-Aminopyridine-2-carboxamide

  • MDL number MFCD06658329
  • Cas Number : 50608-99-6
  • Molecular Weight : 137.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-aminopyridine-2-carboxamide.html

Safety Information

signalword Warning
pcodes P210;P261;P264;P271;P280;P302+352;P304+340;P305+351+338;P337+313;P370+378;P403+233;P403+235
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3697
pictograms GHS07
hstatements H227;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart