ADV371425751 AChemBlock

5-Fluoro-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD18254577
  • Cas Number : 1256788-84-7
  • Molecular Weight : 137.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-fluoro-1h-pyrazolo-3-4-b-pyridine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3700
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart