ADV371425758 AChemBlock

2-Methyl-4-nitropyridine-N-oxide

  • MDL number MFCD00014625
  • Cas Number : 5470-66-6
  • Molecular Weight : 154.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form yellow powder
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-4-nitropyridine-n-oxide.html

Safety Information

signalword Warning
pcodes P210;P280;P370+378;P403+235
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3711
pictograms GHS07
hstatements H227
Availability details for
Quantity
  • Availability
  • Add to Cart