ADV371425764 AChemBlock

4-Pyridazinecarboxaldehyde

  • MDL number MFCD06227447
  • Cas Number : 50901-42-3
  • Molecular Weight : 108.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid/gold solid/green yellow solid/brown or grey solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-pyridazinecarboxaldehyde.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2212
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart