ADV371425769 AChemBlock

3-Hydroxypyrazine-2-carboxylic acid

  • MDL number MFCD00235136
  • Cas Number : 20737-42-2
  • Molecular Weight : 140.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-hydroxypyrazine-2-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3727
pictograms GHS07; GHS09
hstatements H302;H410
Availability details for
Quantity
  • Availability
  • Add to Cart