ADV371425777 AChemBlock

2,6-Dichloro-3-bromobenzoic acid

  • MDL number MFCD18432783
  • Cas Number : 80257-12-1
  • Molecular Weight : 269.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-6-dichloro-3-bromobenzoic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3736
pictograms GHS07; GHS09
hstatements H302;H410
Availability details for
Quantity
  • Availability
  • Add to Cart