ADV371425790 AChemBlock

Methyl oxindole-4-carboxylate

  • MDL number MFCD08277245
  • Cas Number : 90924-46-2
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-oxindole-4-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P264;P271;P280;P302+352;P304+340;P305+351+338;P337+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3750
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart