ADV371425795 AChemBlock

1-Boc-3-iodo-5-azaindole

  • MDL number MFCD08234845
  • Cas Number : 877060-48-5
  • Molecular Weight : 344.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-iodo-5-azaindole.html

Safety Information

signalword Danger
pcodes P264;P270;P280;P301+310;P305+351+338;P337+313
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3755
pictograms GHS06
hstatements H301;H319
Availability details for
Quantity
  • Availability
  • Add to Cart