ADV371425796 AChemBlock

Pyrazolo[1,5-a]pyrimidine-3-carbonitrile

  • MDL number MFCD01927799
  • Cas Number : 25939-87-1
  • Molecular Weight : 144.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid/light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/pyrazolo-1-5-a-pyrimidine-3-carbonitrile.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3756
pictograms GHS07; GHS09
hstatements H302;H410
Availability details for
Quantity
  • Availability
  • Add to Cart