ADV371425804 AChemBlock

3-Amino-1-Boc-3-methylpyrrolidine

  • MDL number MFCD12408544
  • Cas Number : 1158758-59-8
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid/colorless or light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-amino-1-boc-3-methylpyrrolidine.html

Safety Information

signalword Danger
pcodes P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3765
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart