ADV371425805 AChemBlock

Ethyl 4-oxo-1,4-dihydropyrrolo[1,2-b]pyridazine-3-carboxylate

  • MDL number MFCD13195447
  • Cas Number : 156335-37-4
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-oxo-1-4-dihydropyrrolo-1-2-b-pyridazine-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3766
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart