ADV371425808 AChemBlock

Pyrrolo[1,2-b]pyridazin-4(1H)-one

  • MDL number MFCD12923493
  • Cas Number : 888720-26-1
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form gray solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/pyrrolo-1-2-b-pyridazin-4-1h-one.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3770
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart