ADV371425809 AChemBlock

7-Boc-2,4-dichloro-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine

  • MDL number MFCD09025732
  • Cas Number : 916420-27-4
  • Molecular Weight : 304.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-boc-2-4-dichloro-5-6-7-8-tetrahydropyrido-3-4-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2750
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart