ADV371425810 AChemBlock

4,4'-Difluoro-hexahydro-1H-azepine hydrochloride

  • MDL number MFCD11870210
  • Cas Number : 1094073-72-9
  • Molecular Weight : 171.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-4-difluoro-hexahydro-1h-azepine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2752
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart