ADV371425811 AChemBlock

4-Methoxy-hexahydro-1H-azepine

  • MDL number MFCD17015864
  • Cas Number : 1071594-49-4
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-methoxy-hexahydro-1h-azepine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2753
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart