ADV371425819 AChemBlock

5-Bromo-4-chlorothieno[2,3-d]pyrimidine

  • MDL number MFCD11042739
  • Cas Number : 814918-95-1
  • Molecular Weight : 249.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-4-chlorothieno-2-3-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2761
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart