ADV371425828 AChemBlock

1-(6-Amino-3,3-dimethylindolin-1-yl)ethanone

  • MDL number MFCD07636699
  • Cas Number : 453562-71-5
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form pale-red solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-6-amino-3-3-dimethylindolin-1-yl-ethanone.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2770
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart