ADV371425833 AChemBlock

5-Amino-6-bromo-2-pyrazinecarboxylic acid

  • MDL number MFCD04039387
  • Cas Number : 887352-34-3
  • Molecular Weight : 218.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-amino-6-bromo-2-pyrazinecarboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2776
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart