ADV371425840 AChemBlock

(S)-Tetrahydrofuran-3-carboxylic acid

  • MDL number MFCD07369984
  • Cas Number : 168395-26-4
  • Molecular Weight : 116.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow liquid/colorless liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-tetrahydrofuran-3-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2785
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart