ADV371425848 AChemBlock

7H-Pyrrolo[2,3-d]pyrimidine-5-carboxylic acid

  • MDL number MFCD17015971
  • Cas Number : 1234616-77-3
  • Molecular Weight : 163.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7h-pyrrolo-2-3-d-pyrimidine-5-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2793
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart