ADV371425854 AChemBlock

1-Boc-3-hydroxy-3-(aminomethyl)azetidine

  • MDL number MFCD17015986
  • Cas Number : 1008526-71-3
  • Molecular Weight : 202.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-hydroxy-3-aminomethyl-azetidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2800
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart