ADV371425863 AChemBlock

2,4-Dichlorofuro[3,2-d]pyrimidine

  • MDL number MFCD11520866
  • Cas Number : 956034-07-4
  • Molecular Weight : 189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-4-dichlorofuro-3-2-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2818
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart