ADV371425872 AChemBlock

4-Hydroxymethyl-2,2-difluoro-1,3-benzodioxole

  • MDL number MFCD06202698
  • Cas Number : 157437-25-7
  • Molecular Weight : 188.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-hydroxymethyl-2-2-difluoro-1-3-benzodioxole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2827
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart