ADV371425876 AChemBlock

2,4-Dichloro-pyrrolo[2,1-f][1,2,4]triazine

  • MDL number MFCD11044885
  • Cas Number : 918538-05-3
  • Molecular Weight : 188.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-4-dichloro-pyrrolo-2-1-f-1-2-4-triazine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2831
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart