ADV371425879 AChemBlock

1-Boc-3-bromo-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD17016052
  • Cas Number : 1234616-46-6
  • Molecular Weight : 298.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-bromo-1h-pyrazolo-3-4-b-pyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2834
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart