ADV371425881 AChemBlock

6-Bromopyrazolo[1,5-a]pyridine-2-carboxylic acid

  • MDL number MFCD11518929
  • Cas Number : 876379-74-7
  • Molecular Weight : 241.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-bromopyrazolo-1-5-a-pyridine-2-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2836
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart