ADV371425882 AChemBlock

Ethyl 6-bromopyrazolo[1,5-a]pyridine-2-carboxylate

  • MDL number MFCD16657731
  • Cas Number : 1176413-18-5
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-6-bromopyrazolo-1-5-a-pyridine-2-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2837
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart