ADV371425890 AChemBlock

Methyl 6-bromo-(1H)-indazole-4-carboxylate

  • MDL number MFCD07781319
  • Cas Number : 885518-49-0
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-bromo-1h-indazole-4-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2845
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart