ADV371425912 AChemBlock

5-Fluoropyrimidine-2-carbonitrile

  • MDL number MFCD11846243
  • Cas Number : 38275-55-7
  • Molecular Weight : 123.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-fluoropyrimidine-2-carbonitrile.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2871
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart