ADV371425919 AChemBlock

1H-Pyrazole-4-carboxaldehyde hydrochloride

  • MDL number MFCD09759205
  • Cas Number : 1197230-88-8
  • Molecular Weight : 132.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrazole-4-carboxaldehyde-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2878
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart