ADV371425921 AChemBlock

1-Methyl-2-cyanoimidazole

  • MDL number MFCD08236846
  • Cas Number : 45515-45-5
  • Molecular Weight : 107.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-2-cyanoimidazole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2880
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart