ADV371425923 AChemBlock

5-Trifluoromethyl-1H-indole-2-carboxylic acid

  • MDL number MFCD03095229
  • Cas Number : 496946-78-2
  • Molecular Weight : 229.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form gray solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-trifluoromethyl-1h-indole-2-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2882
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart