ADV371425927 AChemBlock

5-Fluoropyridine-2-carboxamidine hydrochloride

  • MDL number MFCD12755545
  • Cas Number : 1179362-15-2
  • Molecular Weight : 175.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-fluoropyridine-2-carboxamidine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2886
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart