ADV371425930 AChemBlock

2-Cyclopropyl-1H-imidazole

  • MDL number MFCD19217297
  • Cas Number : 89532-38-7
  • Molecular Weight : 108.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-cyclopropyl-1h-imidazole.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2889
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart