ADV371425931 AChemBlock

2-(trifluoromethyl)-1H-imidazole

  • MDL number MFCD18415320
  • Cas Number : 66675-22-7
  • Molecular Weight : 136.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1h-imidazole-2-trifluoromethyl.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2890
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart